missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ASCC1 Monoclonal Antibody (OTI1E2), TrueMAB™, OriGene
Mouse Monoclonal Antibody
Brand: Origene Technologies TA503720S
This item is not returnable.
View return policy
Description
ASCC1 Monoclonal antibody specifically detects ASCC1 in Human samples. It is validated for Flow Cytometry, Immunocytochemistry, Western BlotSpecifications
| ASCC1 | |
| Monoclonal | |
| 1 mg/mL | |
| PBS with 1% BSA, 50% glycerol and 0.02% sodium azide | |
| Q8IYX1 | |
| Mouse | |
| Affinity chromatography | |
| RUO | |
| 161514 | |
| -20° C, Avoid Freeze/Thaw Cycles | |
| IgG1 |
| Flow Cytometry, Immunocytochemistry, Western Blot | |
| OTI1E2 | |
| Unconjugated | |
| TBC1D21 | |
| 1810015P09Rik; activating signal cointegrator 1 complex subuni 1-like protein; activating signal cointegrator 1 complex subunit 1; AI550520; Asc-1; ASC-1 complex subunit p50; ASC1p50; Ascc1; CGI-18; p50; SMABF2; Trip4 complex subunit p50 | |
| Full length human recombinant protein of human TBC1D21 produced in HEK293T cell. | |
| 30 μL | |
| Primary | |
| Human | |
| Liquid |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction