missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ ASCC2 Polyclonal Antibody
GREENER_CHOICE

Product Code. 17481453 Shop All Thermo Scientific Products
Change view
Click to view available options
Quantity:
100 μL
Unit Size:
100µL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
17481453 100 μL 100µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 17481453 Supplier Invitrogen™ Supplier No. PA5117793

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

ASCC2 Polyclonal Antibody for Western Blot, IP

Plays a role in DNA damage repair as component of the ASCC complex. Recruits ASCC3 and ALKBH3 to sites of DNA damage by binding to polyubiquitinated proteins that have 'Lys-63'-linked polyubiquitin chains (PubMed:29144457). Part of the ASC-1 complex that enhances NF-kappa-B, SRF and AP1 transactivation (PubMed:12077347). [UniProt].
TRUSTED_SUSTAINABILITY

Specifications

Antigen ASCC2
Applications Immunoprecipitation, Western Blot
Classification Polyclonal
Conjugate Unconjugated
Formulation PBS with 0.03% ProClin 300; pH 7
Gene ASCC2
Gene Accession No. Q9H1I8
Gene Alias 1700011I11Rik; 2610034L15Rik; activating signal cointegrator 1 complex subunit 2; AI482016; ASC-1 complex subunit P100; Asc1p100; ASCC2; AW046480; p100; RGD1561422; trip4 complex subunit p100
Gene Symbols ASCC2
Host Species Rabbit
Immunogen E. coli-derived Human ASCC2 fragment.
Purification Method Antigen affinity chromatography, Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 84164
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.