missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ ASCL2 Polyclonal Antibody

Description
Peptide sequence: QALRQHVPHG GASKKLSKVE TLRSAVEYIR ALQRLLAEHD AVRNALAGGL Sequence homology: Cow: 100%; Dog: 100%; Guinea Pig: 100%; Human: 100%; Mouse: 92%; Rabbit: 77%; Rat: 100%; Zebrafish: 77%.
This gene is a member of the basic helix-loop-helix (BHLH) family of transcription factors. It activates transcription by binding to the E box (5'-CANNTG-3'). Dimerization with other BHLH proteins is required for efficient DNA binding. Involved in the determination of the neuronal precursors in the peripheral nervous system and the central nervous system.
Specifications
Specifications
| Antigen | ASCL2 |
| Applications | Western Blot |
| Classification | Polyclonal |
| Concentration | 0.5 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 2% sucrose and 0.09% sodium azide |
| Gene | ASCL2 |
| Gene Accession No. | Q99929 |
| Gene Alias | 2410083I15Rik; achaete scute-like protein 2; achaete-scute complex homolog 2; achaete-scute complex homolog 2 (Drosophila); achaete-scute complex homolog-like 2; achaete-scute complex-like 2; achaete-scute complex-like protein 2; achaete-scute family bHLH transcription factor 2; achaete-scute homolog 2; Ascl2; ASH2; ASH-2; bHLH transcription factor; Mash2; BHLHA45; Class A basic helix-loop-helix protein 45; H M bHLHa45; HASH2; long transient receptor potential-related channel 5; mammalian achaete scute homolog 2; mammalian achaete/scute homologue 2; MASH2; mASH-2 |
| Gene Symbols | ASCL2 |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?