Learn More
ASCL3 Monoclonal Antibody (OTI1C5), TrueMAB™, OriGene
Mouse Monoclonal Antibody
Brand: Origene Technologies TA811864
This item is not returnable.
View return policy
Description
ASCL3 is a protein coding gene, transcriptional repressor, and inhibits myogenesis.Specifications
| ASCL3 | |
| Monoclonal | |
| Unconjugated | |
| ASCL3 | |
| HASH3, SGN1, bHLHa42 | |
| Mouse | |
| Affinity Chromatography | |
| RUO | |
| 56676 | |
| -20° C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| Western Blot | |
| OTI1C5 | |
| PBS with 1% BSA, 50% glycerol and 0.02% sodium azide | |
| Q9NQ33 | |
| ASCL3 | |
| Full length human recombit protein of human Ascl3 produced in HEK293T cell. | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG2a |