missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ASCL3 Monoclonal Antibody (OTI1C5), TrueMAB™, OriGene
Mouse Monoclonal Antibody
Brand: Origene Technologies TA811864S
This item is not returnable.
View return policy
Description
ASCL3 Monoclonal antibody specifically detects ASCL3 in Human samples. It is validated for Western BlotSpecifications
| ASCL3 | |
| Monoclonal | |
| 1 mg/mL | |
| PBS with 1% BSA, 50% glycerol and 0.02% sodium azide | |
| O43736 | |
| Mouse | |
| Affinity chromatography | |
| RUO | |
| 9452 | |
| -20° C, Avoid Freeze/Thaw Cycles | |
| IgG2a |
| Western Blot | |
| OTI1C5 | |
| Unconjugated | |
| ITM2A | |
| achaete-scute complex homolog 3; achaete-scute complex homolog 3 (Drosophila); achaete-scute complex homolog-like 3; achaete-scute family bHLH transcription factor 3; achaete-scute homolog 3; Ascl3; ASH-3; ASH3A; bHLH transcription factor Sgn-1 (Salivary Glands 1); bHLH transcriptional regulator Sgn-1; bHLHa42; class A basic helix-loop-helix protein 42; HASH3; ICRFP703B1614Q5.4; ICRFP703N2430Q5.4; mASH3; mASH-3; Sgn1 | |
| Full length human recombinant protein of human ITM2A produced in E.coli. | |
| 30 μL | |
| Primary | |
| Human | |
| Liquid |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction