missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ ASCT2 Polyclonal Antibody
Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA533027
This item is not returnable.
View return policy
Description
Percent identity with other species by BLAST analysis: Human, Gorilla (100%) Gibbon, Monkey, Marmoset (93%) Pig (80%).
SLC1A5 (Solute carrier family 1 (neutral amino acid transporter), member 5) is a member of the Na(+)-dependent amino acid transporter superfamily. It has a broad substrate specificity, a preference for zwitterionic amino acids, and a sodium-dependence. It accepts as substrates all neutral amino acids, including glutamine, asparagine, and branched-chain and aromatic amino acids, and excludes methylated amino acids, anionic amino acids, and cationic amino acids. It acts as a cell surface receptor for feline endogenous virus RD114, baboon M7 endogenous virus and type D simian retroviruses.
Specifications
| ASCT2 | |
| Polyclonal | |
| Unconjugated | |
| SLC1A5 | |
| AAAT; ASC-like Na(+)-dependent neutral amino acid transporter ASCT2; Asct2; ATB(0); ATB0; ATBO; baboon M7 virus receptor; CAZ-associated structural protein; H4-ASCT2; H4-system ASC-like transporter; insulin-activated amino acid transporter; M7V1; M7VS1; neutral amino acid transporter B; neutral amino acid transporter B(0); R16; RD114 virus receptor; RD114/simian type D retrovirus receptor; RDR; RDRC; SLC1A5; Slc1a7; sodium-dependent neutral amino acid transporter ASCT2; sodium-dependent neutral amino acid transporter type 2; solute carrier family 1 (neutral amino acid transporter), member 5; solute carrier family 1 member 5; solute carrier family 1, member 7 | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 6510 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin) | |
| 0.6 mg/mL | |
| PBS with 0.1% sodium azide | |
| Q15758 | |
| SLC1A5 | |
| Synthetic 15 amino acid peptide from internal region of human SLC1A5. | |
| 50 μg | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction