Learn More
Asenapine hydrochloride, MedChemExpress
Product Code. 16467078
Encompass_Preferred
Click to view available options
Quantity:
10 mg
50 mg
100 mg
Unit Size:
100mg
10mg
50mg
Product Code. 16467078
Brand: Medchem Express HY16567
MedChemExpress Asenapine hydrochloride, an antipsychotic, is a 5-HT (1A, 1B, 2A, 2B, 2C, 5A, 6, 7) and Dopamine (D2, D3, D4) receptor antagonist with Ki values of 0.03-4.0 nM for 5-HT and 1.3, 0.42, 1.1 nM for Dopamine receptor, respectively.
Asenapine hydrochloride, an antipsychotic, is a 5-HT (1A, 1B, 2A, 2B, 2C, 5A, 6, 7) and Dopamine (D2, D3, D4) receptor antagonist with Ki values of 0.03-4.0 nM for 5-HT and 1.3, 0.42, 1.1 nM for Dopamine receptor, respectively.
Specifications
Chemical Name or Material Asenapine hydrochloride
CAS 1412458-61-7
Color White
Purity Grade Notes Research
Molecular Formula C17H17Cl2NO
Quantity 10 mg
Solubility Information H2O : 100 mg/mL (310.34 mM; Need ultrasonic) ∣DMSO : 50 mg/mL (155.17 mM; Need ultrasonic)
SMILES [H]Cl.ClC1=CC2=C(C=C1)OC3=C(C=CC=C3)[C@](C4)([H])[C@]2([H])CN4C
Molecular Weight (g/mol) 322.23
Formula Weight 322.23
Percent Purity 98.47%
Grade Research
Physical Form Solid
For Use With (Application) Neuroscience-Neuromodulation
Show More Show Less
Research purposes only