missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Asenapine, TRC
Click to view available options
Quantity:
10 mg
Unit Size:
10mg
Specifications
Specifications
| Chemical Name or Material | Asenapine |
| CAS | 65576-45-6 |
| Purity Grade Notes | HPLC |
| Molecular Formula | C17 H16 Cl N O |
| Quantity | 10 mg |
| Synonym | (3aR,12bR)-rel-5-Chloro-2,3,3a,12b-tetrahydro-2-methyl-1H-dibenz[2,3:6,7]oxepino[4,5-c]pyrrole, 1H-Dibenz[2,3:6,7]oxepino[4,5-c]pyrrole, 5-chloro-2,3,3a,12b-tetrahydro-2-methyl-, trans-, Asenapine, Saphris, Sycrest |
| InChI Formula | InChI=1S/C17H16ClNO/c1-19-9-14-12-4-2-3-5-16(12)20-17-7-6-11(18)8-13(17)15(14)10-19/h2-8,14-15H,9-10H2,1H3/t14-,15-/m0/s1 |
| SMILES | CN1C[C@@H]2[C@@H](C1)c3cc(Cl)ccc3Oc4ccccc24 |
| IUPAC Name | (2R,6R)-9-chloro-4-methyl-13-oxa-4-azatetracyclo[12.4.0.02,6.07,12]octadeca-1(18),7(12),8,10,14,16-hexaene |
| Molecular Weight (g/mol) | 285.77 |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction