missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ ASGR1 Monoclonal Antibody (8D7), FITC

Mouse Monoclonal Antibody
Brand: Invitrogen™ MA548985
This item is not returnable.
View return policy
Description
The monoclonal antibody 8D7 is cross reactive with human ASGPR.
The asialoglycooprotein receptor (ASGPR) is a transmembrane glycoprotein (42 kDa) which mediates binding, internalization and degradation of extracellular glycoproteins that have exposed terminal galactose residues. The receptor is expressed on the surface of the hepatocytes in a polar manner, i. e. it is present on the sinusoidal and lateral plasma membranes but not on the bile canalicular membrane. The mammalian hepatic ASGPR mediate the endocytosis and degradation of serum proteins from which terminal sialic residues have been removed. The rat asialoglycoprotein receptor (ASGPR) consists of three polypeptide subunits: rat hepatic lectin (RHL)-1, RHL-2 and RHL-3. The monoclonal antibody 8D7 recognizes a subunit-specific epitope of RHL-1 of rat ASGPR. The monoclonal antibody 8D7 is cross reactive with human ASGPR.
Specifications
| ASGR1 | |
| Monoclonal | |
| 100 μg/mL | |
| PBS with 1% BSA and 0.02% sodium azide | |
| P02706 | |
| Asgr1 | |
| Crude rat liver membrane extracts. | |
| 100 μg | |
| Primary | |
| Rat | |
| Antibody | |
| IgG1 |
| Flow Cytometry, Inhibition Assays, Immunohistochemistry (Frozen), Western Blot, Immunocytochemistry | |
| 8D7 | |
| FITC | |
| Asgr1 | |
| ASGPR; ASGPR 1; ASGP-R 1; ASGPR1; ASGR; ASGR1; Asgr-1; asialoglycoprotein receptor (RHL1); asialoglycoprotein receptor 1; Asialoglycoprotein receptor 1 (hepatic lectin); asialoglycoprotein receptor RHL1; CLEC4H1; C-type lectin domain family 4 member H1; Hepatic lectin 1; hepatic lectin 1 RHL1; hepatic lectin 1, RHL1; hepatic lectin H1; HL-1; RATRHL1; RHL1; RHL-1; Unknown (protein for MGC:133691) | |
| Mouse | |
| Protein G | |
| RUO | |
| 24210 | |
| 4°C, store in dark | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction