missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ASH2L Monoclonal Antibody (6F6B9)

ASH2L Monoclonal Antibody (6F6B9)
Brand: Thermo Scientific MA538447
This item is not returnable.
View return policy
Description
ASH2L Monoclonal Antibody for Western Blot, IHC (P), Flow, ELISA
Specifications
| ASH2L | |
| Monoclonal | |
| 1 mg/mL | |
| PBS with 0.05% sodium azide | |
| Q91X20, Q9UBL3 | |
| ASH2L | |
| Purified recombinant fragment of human ASH2L (aa: 493-628) expressed in E. Coli. | |
| 100 μg | |
| Primary | |
| Human, Mouse | |
| Antibody | |
| IgG1 |
| ELISA, Flow Cytometry, Immunohistochemistry (Paraffin), Western Blot | |
| 6F6B9 | |
| Unconjugated | |
| ASH2L | |
| ASH 2; ASH2; ash2 (absent, small, or homeotic)-like; ash2 (absent, small, or homeotic)-like (Drosophila); ASH2 LIKE; ASH2 like histone lysine methyltransferase complex subunit; ASH2 like protein; ASH2 like, histone lysine methyltransferase complex subunit; Ash2l; ASH2L1; ASH2L2; ash2-like; ASH2-like protein; Bre 2; Bre2; set1/Ash2 histone methyltransferase complex subunit ASH2 | |
| Mouse | |
| Protein G | |
| RUO | |
| 23808, 9070 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction