missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ASH2L Rabbit anti-Human, Mouse, Non-human primate, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 12331-1-AP-20UL
This item is not returnable.
View return policy
Description
The human ASH2L gene encodes a 628 amino acid protein known as ASH2L1, or isoform 1, which contains a nuclear localization signal and PHD finger motif, suggesting that the gene product functions as a transcription regulator. Alternative splicing results in a shorter isoform 2, designated ASH2L2, which is missing the first 94 amino acid residues found in ASH2L1. Human ASH2L proteins are 60% homologous to Drosophila ash2, which positively regulates expression of certain genes in early development, and contain similar, but not identical, domains, including a zinc finger motif. ASH2L is highly expressed in fetal liver, testis and leukemia cell lines with erythroid and megakaryocytic potential, such as K562, Hel and Dami. Differentiation inducers (e.g. phorbol ester and hemin) cause different expression patterns in these cells lines, suggesting that ASH2L plays a role in hematopoiesis and is associated with particular types of leukemia.Specifications
| ASH2L | |
| Polyclonal | |
| Unconjugated | |
| ASH2L | |
| ASH2, ASH2 like protein, ASH2L, ASH2L1, ASH2L2, Bre2 | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 23808, 290829, 9070 | |
| -20°C | |
| Liquid |
| Western Blot, Immunoprecipitation, Immunohistochemistry (Paraffin) | |
| 0.27 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| Q91X20, Q9UBL3 | |
| Ash2l | |
| ASH2L Fusion Protein Ag3008 | |
| 20 μL | |
| Primary | |
| Human, Non-human Primate, Rat, Mouse | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction