missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Asialoglycoprotein Receptor 2 Antibody (2327C) [Biotin], Novus Biologicals™
Rabbit Monoclonal Antibody
Brand: Novus Biologicals FAB9970B
This item is not returnable.
View return policy
Description
Asialoglycoprotein Receptor 2 Monoclonal antibody specifically detects Asialoglycoprotein Receptor 2 in Human samples. It is validated for Western Blot, Flow Cytometry, Immunohistochemistry
Specifications
| Asialoglycoprotein Receptor 2 | |
| Monoclonal | |
| Biotin | |
| PBS | |
| Rabbit | |
| Protein A or G purified from hybridoma culture supernatant | |
| RUO | |
| Primary | |
| Human | |
| Purified |
| Western Blot, Flow Cytometry, Immunohistochemistry | |
| 2327C | |
| Western Blot, Flow Cytometry, Immunohistochemistry | |
| ASGPR 2, ASGP-R 2, ASGPR2, ASGP-R2, asialoglycoprotein receptor 2, CLEC4H2FLJ60040, C-type lectin domain family 4 member H2, HBxAg-binding protein, HBXBP, Hepatic lectin H2, HL-2 | |
| Chinese hamster ovary cell line CHO-derived recombinant human ASGR2, Gln80-Ala311, Accession # P07307 | |
| 0.1 mL | |
| Signal Transduction | |
| 433 | |
| Store at 4°C in the dark. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction