missing translation for 'onlineSavingsMsg'
Learn More

USP10 Antibody, DyLight 755, Novus Biologicals™

Product Code. 18460473 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18460473 0.1 mL 0.1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18460473 Supplier Novus Biologicals Supplier No. NBP177170IR

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

USP10 Polyclonal antibody specifically detects USP10 in Human, Mouse samples. It is validated for Western Blot, ELISA, Immunofluorescence
TRUSTED_SUSTAINABILITY

Specifications

Antigen USP10
Applications Western Blot, ELISA, Immunofluorescence
Classification Polyclonal
Conjugate DyLight 755
Formulation 50mM Sodium Borate
Gene Alias Deubiquitinating enzyme 10, EC 3.4.19.12, KIAA0190MGC2621, ubiquitin carboxyl-terminal hydrolase 10, ubiquitin specific peptidase 10, ubiquitin specific protease 10, ubiquitin thioesterase 10, Ubiquitin thiolesterase 10, Ubiquitin-specific-processing protease 10, UBPO
Host Species Rabbit
Immunogen Antibody was raised against a 16 amino acid synthetic peptide near the amino terminus of human USP10. The immunogen is located within amino acids 30 - 80 of USP10. Amino Acid Squence: CVDKLPDGQEYQRIEFG
Purification Method Peptide affinity purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Cell Biology
Primary or Secondary Primary
Gene ID (Entrez) 9100
Target Species Human, Mouse
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.