missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ ASIC3 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA599026
This item is not returnable.
View return policy
Description
ASIC3 Polyclonal Antibody for Western Blot, IHC (P), ELISA
ACCN3 encodes a member of the degenerin/epithelial sodium channel (DEG/ENaC) superfamily. The members of this family are amiloride-sensitive sodium channels that contain intracellular N and C termini, two hydrophobic transmembrane regions, and a large extracellular loop, which has many cysteine residues with conserved spacing. ACCN3 encodes a member that is an acid sensor and may play an important role in the detection of lasting pH changes. In addition, a heteromeric association between this member and ACCN1 has been observed as proton-gated channels sensitive to gadolinium.
Specifications
| ASIC3 | |
| Polyclonal | |
| Unconjugated | |
| ASIC3 | |
| ACCN3; acid sensing (proton gated) ion channel 3; acid sensing ion channel 3; acid sensing ion channel subunit 3; acid-sensing (proton-gated) ion channel 3; acid-sensing ion channel 3; Amiloride-sensitive cation channel 3; amiloride-sensitive cation channel 3, testis; Asic3; AW742291; dorsal root ASIC; DRASIC; hASIC3; hTNaC1; modulatory subunit of ASIC2a; neuronal amiloride-sensitive cation channel 3; proton gated cation channel DRASIC; proton-gated cation channel subunit; SLNAC1; Testis sodium channel 1; TNaC1 | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 9311 | |
| -20°C or -80°C if preferred | |
| Liquid |
| ELISA, Immunohistochemistry (Paraffin), Western Blot | |
| 0.6 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| Q9UHC3 | |
| ASIC3 | |
| Recombinant Human Acid-sensing ion channel 3 protein (62-320AA). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction