missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ASMTL Antibody, DyLight 755, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-97916IR
This item is not returnable.
View return policy
Description
ASMTL Polyclonal specifically detects ASMTL in Human samples. It is validated for Western Blot.
Specifications
| ASMTL | |
| Polyclonal | |
| Western Blot | |
| acetylserotonin N-methyltransferase-like, acetylserotonin O-methyltransferase-like, ASMTLX, ASMTLY, ASTML, N-acetylserotonin O-methyltransferase-like protein | |
| Produced in rabbits immunized with E. coli-derived Human ASMTL fragment. | |
| 0.1 mL | |
| Stem Cell Markers | |
| 8623 | |
| Store at 4°C in the dark. | |
| IgG |
| Western Blot | |
| DyLight 755 | |
| 50mM Sodium Borate | |
| Rabbit | |
| Antigen Affinity-purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction