missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ASMTL, Rabbit anti-Human, Polyclonal Antibody, Abnova™
Rabbit Polyclonal Antibody
Brand: Abnova H00008623-D01P.100ug
This item is not returnable.
View return policy
Description
Sequence: MVLCPVIGKLLHKRVVLASASPRRQEILSNAGLRFEVVPSKFKEKLDKASFATPYGYAMETAKQKALEVANRLYQKDLRAPDVVIGADTIVTVGGLILEKPVDKQDAYRMLSRLSGREHSVFTGVAIVHCSSKDHQLDTRVSEFYEETKVKFSELSEELLWEYVHSGEPMDKAGGYGIQALGGMLVESVHGDFLNVVGFPLNHFCKQLVKLYYPPRPEDLRRSVKHDSIPAADTFEDLSDVEGGGSEPTQRDAGSRDEKAEAGEAGQATAEAECHRTRETLPPFPTRLLELIEGFMLSKGLLTACKLKVFDLLKDEAPQKAADIASKVDASACGMERLLDICAAMGLLEKTEQGYSNTETANVYLASDGEYSLHGFIMHNNDLTWNLFTYLEFAIREGTNQHHRALGKKAEDLFQDAYYQSPETRLRFMRAMHGMTKLTACQVATAFNLSRFSSACDVGGCTGALARELAREYPRMQVTVFDLPDIIELAAHFQPPGPQAVQIHFAAGDFFRDPLPSAELYVLCRILHDWPDDKVHKLLSKVAESCKPGAGLLLVETLLDEEKRVAQRALMQSLNMLVQTEGKERSLGEYQCLLELHGFHQVQVVHLGGVLDAILATKVAPSpecifications
| ASMTL | |
| Polyclonal | |
| In 1x PBS, pH 7.4 | |
| BC002508 | |
| ASMTL | |
| ASMTL (AAH02508.1, 1 a.a. ∼ 621 a.a) full-length human protein. | |
| RUO | |
| 8623 | |
| Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. | |
| Purified |
| Western Blot | |
| Unconjugated | |
| acetylserotonin O-methyltransferase-like | |
| ASMTLX/ASMTLY/ASTML | |
| Rabbit | |
| 100 μg | |
| Primary | |
| Human | |
| Antibody |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction