missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ASNA1 Antibody (2A1), DyLight 680, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-42661FR
This item is not returnable.
View return policy
Description
ASNA1 Monoclonal specifically detects ASNA1 in Human samples. It is validated for Western Blot, Flow Cytometry, ELISA, Immunocytochemistry/Immunofluorescence.
Specifications
| ASNA1 | |
| Monoclonal | |
| DyLight 680 | |
| 50mM Sodium Borate with 0.05% Sodium Azide | |
| GET3 | |
| Recombinant human ASNA1 (1-348aa) purified from E. coli | |
| 100 μL | |
| metabolism | |
| 439 | |
| Store at 4C in the dark. | |
| IgG2b κ |
| Western Blot, Flow Cytometry, ELISA, Immunocytochemistry, Immunofluorescence | |
| 2A1 | |
| Western Blot, Flow Cytometry, ELISA, Immunocytochemistry/Immunofluorescence | |
| ARSA, arsA arsenite transporter, ATP-binding, homolog 1 (bacterial), ARSA1, ARSA-I, Arsenical pump-driving ATPase, Arsenite-stimulated ATPase, ATPase ASNA1, EC 3.6, EC 3.6.3.16, GET3, golgi to ER traffic 3 homolog, hARSA-I, hASNA-I, MGC3821, Transmembrane domain recognition complex 40 kDa ATPase subunit, transmembrane domain recognition complex, 40kDa, TRC40arsA (bacterial) arsenite transporter, ATP-binding, homolog 1 | |
| Mouse | |
| Protein A purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction