missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ASNS Monoclonal Antibody (OTI1A10), TrueMAB™, OriGene
Mouse Monoclonal Antibody
Brand: Origene Technologies TA503452S
This item is not returnable.
View return policy
Description
ASNS Monoclonal antibody specifically detects ASNS in Human samples. It is validated for Immunocytochemistry, Western BlotSpecifications
| ASNS | |
| Monoclonal | |
| 1 mg/mL | |
| PBS with 1% BSA, 50% glycerol and 0.02% sodium azide | |
| P37268 | |
| Mouse | |
| Affinity chromatography | |
| RUO | |
| 2222 | |
| -20° C, Avoid Freeze/Thaw Cycles | |
| IgG2b |
| Immunocytochemistry, Western Blot | |
| OTI1A10 | |
| Unconjugated | |
| FDFT1 | |
| ASNS; ASNSD; asparagine synthetase; asparagine synthetase (glutamine-hydrolyzing); asparagine synthetase [glutamine-hydrolyzing]; Cell cycle control protein TS11; Glutamine-dependent asparagine synthetase; TS11; TS11 cell cycle control protein | |
| Full length human recombinant protein of human FDFT1 produced in HEK293T cell. | |
| 30 μL | |
| Primary | |
| Human | |
| Liquid |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction