missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ASPA Antibody, DyLight 650, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-97185C
This item is not returnable.
View return policy
Description
ASPA Polyclonal antibody specifically detects ASPA in Human samples. It is validated for Immunohistochemistry (Paraffin)
Specifications
| ASPA | |
| Polyclonal | |
| 50mM Sodium Borate | |
| Rabbit | |
| Antigen and protein A Affinity-purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
| Immunohistochemistry (Paraffin) | |
| DyLight 650 | |
| ACY-2, ACY2aminoacylase 2, Aminoacylase-2, ASPaminoacylase-2, aspartoacylase, aspartoacylase (aminoacylase 2, Canavan disease), EC 3.5.1.15 | |
| Produced in rabbits immunized with E. coli-derived Human ASPA fragment. | |
| 0.1 mL | |
| Neuroscience | |
| 443 | |
| Store at 4°C in the dark. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction