missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ASPA Monoclonal Antibody (OTI3B5), TrueMAB™, OriGene
Mouse Monoclonal Antibody
Brand: Origene Technologies TA805343
This item is not returnable.
View return policy
Description
ASPA is an enzyme that catalyzes the conversion of N-acetyl_L-aspartic acid (NAA) to aspartate and acetate. NAA is abundant in the brain where hydrolysis by aspartoacylase is thought to help maintain white matter. This protein is an NAA scavenger in other tissues.Specifications
| ASPA | |
| Monoclonal | |
| 1 mg/mL | |
| PBS with 1% BSA, 50% glycerol and 0.02% sodium azide | |
| P45381 | |
| ASPA | |
| Human recombit protein fragment corresponding to amino acids 77-313 of human ASPA produced in E.coli. | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG2a |
| Western Blot | |
| OTI3B5 | |
| Unconjugated | |
| ASPA | |
| ACY2, ASP | |
| Mouse | |
| Affinity Chromatography | |
| RUO | |
| 443 | |
| -20° C, Avoid Freeze/Thaw Cycles | |
| Liquid |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction