Learn More
ROBO4 Monoclonal Antibody (OTI4E2), TrueMAB™, OriGene
Mouse Monoclonal Antibody
Brand:  Origene Technologies TA811323S
Encompass_Preferred
 View more versions of this product
Product Code. 17545244
  • € 242.00 / 30µL
Description
ROBO4 Monoclonal antibody specifically detects ROBO4 in Human samples. It is validated for Western Blot
Specifications
Specifications
ROBO4
Monoclonal
1 mg/mL
PBS with 1% BSA, 50% glycerol and 0.02% sodium azide
P49640
Mouse
Affinity chromatography
RUO
2128
-20° C, Avoid Freeze/Thaw Cycles
IgG1
Western Blot
OTI4E2
Unconjugated
EVX1
1200012D01Rik; AI593217; ECSM4; FLJ20798; Magic roundabout; MGC133352; MGC133353; MRB; ROBO4; roundabout guidance receptor 4; roundabout homolog 4; roundabout homolog 4 (Drosophila); roundabout homolog 4, magic roundabout; roundabout, axon guidance receptor, homolog 4; roundabout, axon guidance receptor, homolog 4 (Drosophila); UNQ421/PRO3674
Human recombinant protein fragment corresponding to amino acids 1-244 of human EVX1 produced in E.coli.