missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Asparagine Synthetase Recombinant Rabbit Monoclonal Antibody (23GB1475), Invitrogen™

Rabbit Recombinant Monoclonal Antibody
€ 186.00 - € 446.00
Specifications
| Antigen | Asparagine Synthetase |
|---|---|
| Clone | 23GB1475 |
| Applications | Flow Cytometry, Western Blot, Immunocytochemistry |
| Classification | Recombinant Monoclonal |
| Conjugate | Unconjugated |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Produktcode | Marke | Quantity | Preis | Menge & Verfügbarkeit | |||||
30087817
![]() |
Invitrogen™
MA553170 |
100 μL |
€ 446.00
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
30088175
|
Invitrogen™
MA553171 |
20 μL |
€ 186.00
20µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Beschreibung
Asparagine Synthetase Recombinant Monoclonal antibody specifically detects Asparagine Synthetase in Human, Mouse, Rat samples. It is validated for Western BlotSpezifikation
| Asparagine Synthetase | |
| Flow Cytometry, Western Blot, Immunocytochemistry | |
| Unconjugated | |
| Rabbit | |
| Human, Mouse, Rat | |
| P08243, P49088, Q61024 | |
| 25612, 27053, 440 | |
| IgG | |
| Affinity chromatography | |
| Antibody |
| 23GB1475 | |
| Recombinant Monoclonal | |
| Liquid | |
| RUO | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
| ASNS; ASNSD; asparagine synthetase; asparagine synthetase (glutamine-hydrolyzing); asparagine synthetase [glutamine-hydrolyzing]; Cell cycle control protein TS11; Glutamine-dependent asparagine synthetase; TS11; TS11 cell cycle control protein | |
| A synthesized peptide derived from human ASNS (500-561AA). | |
| Primary | |
| -20°C | |
| Asns |
Haben Sie Verbesserungsvorschläge?Übermitteln Sie eine inhaltliche Korrektur
Berichtigung von Produktinhalten
Bitte geben Sie uns Ihr Feedback zu den Produktinhalten, indem Sie das folgende Formular ausfüllen.
Name des Produkts