missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Aspartate Aminotransferase Antibody (SR1790), Novus Biologicals™
Rabbit Monoclonal Antibody
€ 349.00 - € 449.00
Specifications
| Antigen | Aspartate Aminotransferase |
|---|---|
| Clone | SR1790 |
| Dilution | Western Blot 1:500-1:2000, Flow Cytometry 1:50, Immunohistochemistry 1:50-1:200, Immunocytochemistry/Immunofluorescence 1:50-1:200 |
| Applications | Western Blot, Flow Cytometry, Immunohistochemistry, Immunofluorescence |
| Classification | Monoclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18651394
|
Novus Biologicals
NBP3-21842-100ul |
100 μg |
€ 449.00
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18652625
|
Novus Biologicals
NBP3-21842-50ul |
50 μg |
€ 349.00
50µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Aspartate Aminotransferase Monoclonal antibody specifically detects Aspartate Aminotransferase in Human, Mouse, Rat samples. It is validated for Western Blot, Flow Cytometry, Immunohistochemistry, ImmunofluorescenceSpecifications
| Aspartate Aminotransferase | |
| Western Blot 1:500-1:2000, Flow Cytometry 1:50, Immunohistochemistry 1:50-1:200, Immunocytochemistry/Immunofluorescence 1:50-1:200 | |
| Monoclonal | |
| Purified | |
| RUO | |
| Human, Mouse, Rat | |
| aspartate aminotransferase, cytoplasmic, ASTQTL1, CASPAT, CCAT, Cysteine aminotransferase, cytoplasmic, Cysteine transaminase, cytoplasmic, EC 2.6.1.1, EC 2.6.1.3, GIG18, Glutamate oxaloacetate transaminase 1, glutamic-oxaloacetic transaminase 1, soluble (aspartate aminotransferase 1), GOT1, growth-inhibiting protein 18, Transaminase A | |
| A synthesized peptide derived from human Aspartate Aminotransferase (Uniprot #: P17174) | |
| Primary | |
| Store at -20° C. Avoid freeze/thaw cycles. |
| SR1790 | |
| Western Blot, Flow Cytometry, Immunohistochemistry, Immunofluorescence | |
| Unconjugated | |
| Rabbit | |
| Lipid and Metabolism | |
| PBS, pH 7.4, 150mM NaCl, 50% glycerol. | |
| 2805 | |
| IgG | |
| Affinity purified |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title