missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Aspartoacylase Monoclonal Antibody (OTI2D2)
GREENER_CHOICE

Product Code. 15916324
Change view
Click to view available options
Quantity:
100 μL
Unit Size:
100µL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15916324 100 μL 100µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 15916324 Supplier Invitrogen™ Supplier No. MA526728

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Monoclonal Antibody

Aspartoacylase Monoclonal Antibody for Western Blot, IHC (P)

ASPA is an enzyme that catalyzes the conversion of N-acetyl_L-aspartic acid (NAA) to aspartate and acetate. NAA is abundant in the brain where hydrolysis by aspartoacylase is thought to help maintain white matter. This protein is an NAA scavenger in other tissues.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Aspartoacylase
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Monoclonal
Clone OTI2D2
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 1% BSA, 50% glycerol and 0.02% sodium azide; pH 7.3
Gene ASPA
Gene Accession No. P45381
Gene Alias ACY2; ACY-2; aminoacylase 2; aminoacylase-2; ASP; ASPA; Aspartoacylase; aspartoacylase (aminoacylase) 2; nur7
Gene Symbols ASPA
Host Species Mouse
Immunogen Human recombinant protein fragment corresponding to amino acids 77-313 of ASPA produced in E.coli.
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 443
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG2a
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.