missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Aspartoacylase Monoclonal Antibody (OTI5B8)

Mouse Monoclonal Antibody
Brand: Invitrogen™ MA526717
This item is not returnable.
View return policy
Description
Aspartoacylase Monoclonal Antibody for Western Blot, IHC (P)
ASPA is an enzyme that catalyzes the conversion of N-acetyl_L-aspartic acid (NAA) to aspartate and acetate. NAA is abundant in the brain where hydrolysis by aspartoacylase is thought to help maintain white matter. This protein is an NAA scavenger in other tissues.
Specifications
| Aspartoacylase | |
| Monoclonal | |
| 1 mg/mL | |
| PBS with 1% BSA, 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| P45381 | |
| ASPA | |
| Human recombinant protein fragment corresponding to amino acids 77-313 of ASPA produced in E.coli. | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG1 |
| Immunohistochemistry (Paraffin), Western Blot | |
| OTI5B8 | |
| Unconjugated | |
| ASPA | |
| ACY2; ACY-2; aminoacylase 2; aminoacylase-2; ASP; ASPA; Aspartoacylase; aspartoacylase (aminoacylase) 2; nur7 | |
| Mouse | |
| Affinity chromatography | |
| RUO | |
| 443 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction