missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ASPDH Antibody (OTI4G8), Alexa Fluor™ 594, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-72508AF594
This item is not returnable.
View return policy
Description
ASPDH Monoclonal specifically detects ASPDH in Human, Mouse, Rat samples. It is validated for Immunohistochemistry.
Specifications
| ASPDH | |
| Monoclonal | |
| Alexa Fluor 594 | |
| 50 mM sodium borate with 0.05% sodium azide | |
| ASPDH | |
| Full length recombinant protein of human ASPDH (NP_001019827) produced in E.coli. | |
| 0.1 mL | |
| 554235.0 | |
| Store at 4°C in the dark. |
| Immunohistochemistry | |
| OTI4G8 | |
| Immunohistochemistry | |
| aspartate dehydrogenase domain containing, Aspartate dehydrogenase domain-containing protein, EC 1.4.1.21, putative L-aspartate dehydrogenase | |
| Mouse | |
| Affinity Purified | |
| Primary | |
| Human, Mouse, Rat | |
| IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction