missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ASPDH Antibody (OTI4G8) - Azide and BSA Free, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-72508
This item is not returnable.
View return policy
Description
ASPDH Monoclonal specifically detects ASPDH in Human, Mouse, Rat samples. It is validated for Immunohistochemistry, Immunohistochemistry-Paraffin.
Specifications
| ASPDH | |
| Monoclonal | |
| LYOPH | |
| Immunohistochemistry 1:150 | |
| aspartate dehydrogenase domain containing, Aspartate dehydrogenase domain-containing protein, EC 1.4.1.21, putative L-aspartate dehydrogenase | |
| Mouse | |
| 18.5 kDa | |
| 100 μg | |
| 554235 | |
| Human, Mouse, Rat | |
| Lyophilized |
| Immunohistochemistry | |
| OTI4G8 | |
| Unconjugated | |
| Lyophilized from PBS (pH 7.3) with 8% Trehalose with No Preservative | |
| ASPDH | |
| Full length human recombinant protein of human ASPDH (NP_001019827) produced in E.coli. | |
| Affinity Purified | |
| Primary | |
| Reconstitute with PBS (pH 7.3). To use this carrier-free antibody for conjugation experiments, another round of desalting is highly recommended. | |
| Store at -20°C. Avoid freeze-thaw cycles. | |
| IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction