Learn More
Aspergillus Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NB600-1374
This item is not returnable.
View return policy
Description
Aspergillus Polyclonal specifically detects Aspergillus in Fungi samples. It is validated for ELISA, Immunohistochemistry, Immunohistochemistry-Paraffin, Immunohistochemistry-Frozen, Immunofluorescence.
Specifications
| ELISA, Immunohistochemistry, Immunohistochemistry (Paraffin), Immunohistochemistry (Frozen) | |
| 0.1 mL | |
| IgG |
| Soluble extract from A. fumigatus, A. flavus, A. niger and A. terreus. | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |