missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Aspergillus Polyclonal Antibody, FITC
Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA173164
This item is not returnable.
View return policy
Description
The PA1-73164 antibody reacts with Aspergillus species. PA1-73164 has been successfully used in Immunofluorescence applications. The PA1-73164 antibody was raised against soluble extract from A. fumigatus, A. flavus, A. niger and A. terreus.
Aspergillus species are highly aerobic and are found in almost all oxygen-rich environments, where they commonly grow as molds on the surface of a substrate, as a result of the high oxygen tension. Some Aspergillus species cause serious disease in humans.
Specifications
| Aspergillus | |
| Polyclonal | |
| FITC | |
| Rabbit | |
| IgG fraction | |
| RUO | |
| Fungi | |
| Antibody | |
| IgG |
| Immunocytochemistry | |
| 5.0 mg/mL | |
| PBS with 10mg/mL BSA and 0.1% sodium azide; pH 7.2 | |
| Soluble extract from A. fumigatus, A. flavus, A. niger and A. terreus. | |
| 1 mL | |
| Primary | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. Store in the dark. | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction