missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ ASPM Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA599790
This item is not returnable.
View return policy
Description
Antibody detects endogenous levels of total ASPM.
This gene is the human ortholog of the Drosophila melanogaster 'abnormal spindle' gene (asp), which is essential for normal mitotic spindle function in embryonic neuroblasts. Studies in mouse also suggest a role of this gene in mitotic spindle regulation, with a preferential role in regulating neurogenesis. Mutations in this gene are associated with microcephaly primary type 5. Multiple transcript variants encoding different isoforms have been found for this gene.
Specifications
| ASPM | |
| Polyclonal | |
| Unconjugated | |
| ASPM | |
| abnormal spindle microtubule assembly; Abnormal spindle protein homolog; Abnormal spindle-like microcephaly-associated protein; ASP; asp (abnormal spindle) homolog, microcephaly associated; Asp homolog; ASPM; Calmbp1; MCPH5 | |
| Rabbit | |
| Affinity Chromatography | |
| RUO | |
| 259266, 289054 | |
| -20°C | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry | |
| 1 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
| Q8IZT6 | |
| ASPM | |
| A synthesized peptide derived from human ASPM(Accession Q8IZT6), corresponding to amino acid residues L1261-R1311. | |
| 100 μL | |
| Primary | |
| Human, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction