missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Asporin Polyclonal Antibody
GREENER_CHOICE

Product Code. 16556191
Change view
Click to view available options
Quantity:
100 μL
Unit Size:
100µL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16556191 100 μL 100µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 16556191 Supplier Invitrogen™ Supplier No. PA587799

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

Immunogen sequence: PFDTRMLDLQ NNKIKEIKEN DFKGLTSLYG LILNNNKLTK IHPKAFLTTK KLRRLYLSHN QLSEIPLNLP KSLAELRIHE NKVKKIQKDT FKGMNALHVL EMSANPLDNN GIEPGAFEGV TVFHIRIAEA KLTSVPKGLP PTLLELHLDY NKISTVELED FKRYKELQRL GLGNNKITDI ENGSLANIPR VREIHLENNK LKKIPSGLPE LKYLQIIFLH SNSIARVGVN DFCPTVPKMK KSLYSAISLF NNPVKYWEMQ PATFRCVLSR MSVQLGNFGM; Positive Samples: HeLa, SKOV3, SGC-7901, Mouse heart, Mouse stomach, Rat heart; Cellular Location: extracellular matrix, extracellular space.

ASPN belongs to a family of leucine-rich repeat (LRR) proteins associated with the cartilage matrix. The name asporin reflects the unique aspartate-rich N terminus and the overall similarity to decorin.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Asporin
Applications ELISA, Western Blot
Classification Polyclonal
Concentration 2.56 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
Gene Aspn
Gene Accession No. Q99MQ4, Q9BXN1
Gene Alias 4631401G09Rik; AA986886; ASPN; Asporin; asporin (LRR class 1); asporin proteoglycan; FLJ20129; OS3; periodontal ligament associated protein 1; periodontal ligament-associated protein 1; PLAP1; PLAP-1; SLRR1C; small leucine-rich protein 1C; UNQ215/PRO241
Gene Symbols Aspn
Host Species Rabbit
Immunogen Recombinant fusion protein containing a sequence corresponding to amino acids 100-379 of human ASPN (NP_0601504).
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 306805, 54829, 66695
Target Species Human, Mouse, Rat
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.