missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ASPP1 Antibody, Alexa Fluor™ 532, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NB110-40683AF532
This item is not returnable.
View return policy
Description
ASPP1 Polyclonal antibody specifically detects ASPP1 in Human samples. It is validated for Western Blot
Specifications
| ASPP1 | |
| Polyclonal | |
| Western Blot | |
| apoptosis-stimulating protein of p53, 1, ASPP1Protein phosphatase 1 regulatory subunit 13B, KIAA0771apoptosis-stimulating of p53 protein 1, p53BP2-like, p85, protein phosphatase 1, regulatory (inhibitor) subunit 13B | |
| The immunogen recognized by this antibody maps to a region between residue 50 and 100 of human Apoptosis-Stimulated Protein of p53, 1 using the numbering given in entry NP_056131.1 (GeneID 23368). | |
| 0.1 mL | |
| Primary | |
| Human | |
| Purified |
| Western Blot | |
| Alexa Fluor 532 | |
| 50mM Sodium Borate | |
| Rabbit | |
| Immunogen affinity purified | |
| RUO | |
| 23368 | |
| Store at 4C in the dark. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction