Learn More
ASRGL1 Antibody (CL1679) [mFluor Violet 610 SE], Novus Biologicals™
Mouse Monoclonal Antibody
Brand:  Novus Biologicals NBP2-42378MFV610
Product Code. 30292329
  • € 780.00 / 0.10mL
Description
ASRGL1 Monoclonal antibody specifically detects ASRGL1 in Human samples. It is validated for Immunohistochemistry,Western Blot,Immunohistochemistry (Paraffin)
TRUSTED_SUSTAINABILITY
Specifications
Specifications
ASRGL1
Monoclonal
mFluor Violet 610 SE
ALP1, ALPL-asparagine amidohydrolase, asparaginase like 1, asparaginase-like 1 protein, Asparaginase-like protein 1, CRASH, EC 3.5.1.1, FLJ22316, L-asparaginase
This antibody was developed using a recombinant protein derived from Q7L266, with the exact immunogen sequence remaining proprietary.
0.1 mL
Neuroscience
80150
Store at 4⪚C in the dark.
IgG1
Immunohistochemistry, Western Blot, Immunohistochemistry (Paraffin)
CL1679
50mM Sodium Borate
Mouse
Protein A purified