missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ASRGL1 Antibody (CRASH/1290), DyLight 755, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP3-08327IR
This item is not returnable.
View return policy
Description
ASRGL1 Monoclonal antibody specifically detects ASRGL1 in Human samples. It is validated for ELISA
Specifications
| ASRGL1 | |
| Monoclonal | |
| DyLight 755 | |
| ALP1, ALPL-asparagine amidohydrolase, asparaginase like 1, asparaginase-like 1 protein, Asparaginase-like protein 1, CRASH, EC 3.5.1.1, FLJ22316, L-asparaginase | |
| Recombinant human ASRGL1 fragment (Uniprot: Q7L266) | |
| 100 μg | |
| Neuroscience | |
| 80150 | |
| Store at 4°C in the dark. | |
| IgG1 κ |
| ELISA | |
| CRASH/1290 | |
| 50mM Sodium Borate | |
| Mouse | |
| Protein A or G purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction