missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ASS1 Monoclonal Antibody (OTI3D1), TrueMAB™, OriGene
Mouse Monoclonal Antibody
Brand: Origene Technologies TA809168S
This item is not returnable.
View return policy
Description
ASS1 Monoclonal antibody specifically detects ASS1 in Human samples. It is validated for Immunohistochemistry (Paraffin), Western BlotSpecifications
| ASS1 | |
| Monoclonal | |
| 1 mg/mL | |
| PBS with 1% BSA, 50% glycerol and 0.02% sodium azide | |
| Q96EB1 | |
| Mouse | |
| Affinity chromatography | |
| RUO | |
| 26610 | |
| -20° C, Avoid Freeze/Thaw Cycles | |
| IgG1 |
| Immunohistochemistry (Paraffin), Western Blot | |
| OTI3D1 | |
| Unconjugated | |
| Elp4 | |
| AA408052; arginino succinate synthetase; argininosuccinate synthase; argininosuccinate synthase 1; argininosuccinate synthetase; argininosuccinate synthetase 1; arginosuccinate synthetase 1; ASS; Ass1; Ass-1; ASSA; citrulline-aspartate ligase; citrulline--aspartate ligase; citrullinemia; CN; CTLN1; fold; mutant arginino succinate synthetase; RP11-618A20.2 | |
| Human recombinant protein fragment corresponding to amino acids 133-407 of human ELP4 produced in E.coli. | |
| 30 μL | |
| Primary | |
| Human | |
| Liquid |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction