missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ ASS1 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA572474
This item is not returnable.
View return policy
Description
ASS1 Polyclonal Antibody for Western Blot, ICC/IF, IHC (P), Flow
The protein encoded by this gene catalyzes the penultimate step of the arginine biosynthetic pathway. There are approximately 10 to 14 copies of this gene including the pseudogenes scattered across the human genome, among which the one located on chromosome 9 appears to be the only functional gene for argininosuccinate synthetase. Mutations in the chromosome 9 copy of ASS cause citrullinemia. Two transcript variants encoding the same protein have been found for this gene.
Specifications
| ASS1 | |
| Polyclonal | |
| Unconjugated | |
| ASS1 | |
| AA408052; arginino succinate synthetase; argininosuccinate synthase; argininosuccinate synthase 1; argininosuccinate synthetase; argininosuccinate synthetase 1; arginosuccinate synthetase 1; ASS; Ass1; Ass-1; ASSA; citrulline-aspartate ligase; citrulline--aspartate ligase; citrullinemia; CN; CTLN1; fold; mutant arginino succinate synthetase; RP11-618A20.2 | |
| Rabbit | |
| Antigen Affinity Chromatography, Protein A | |
| RUO | |
| 11898, 25698, 445 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Flow Cytometry, Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry | |
| 1.45 mg/mL | |
| PBS with 0.09% sodium azide; pH 7.4 | |
| P00966, P09034, P16460 | |
| ASS1 | |
| KLH conjugated synthetic peptide between 192-221 amino acids from the Central region of human ASS. | |
| 400 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction