Learn More
DiscoverX AssayComplete™ Cell Plating 27 Reagent
Click to view available options
Quantity:
100 mL
Unit Size:
100mL
Description
AssayComplete™ Cell Plating 27 Reagent is an optimized cell plating reagent for use in plating of DiscoverX cell lines for assays. This reagent was optimized for maximal cell health and ligand responsiveness during assays while eliminating hormones and growth components which may lead to increased basal activity. This product is provided as a bottle of 100mL. Please refer to cell line specific datasheet to determine the recommended cell plating reagent.
Specifications
Specifications
| For Use With (Application) | Cell Plating Reagents |
| Quantity | 100 mL |