Learn More
DiscoverX AssayComplete™ Thawing Reagent T8
Product Code. 17392659
Encompass
Click to view available options
Quantity:
100 mL
Unit Size:
100mL
Product Code. 17392659
Brand: DiscoverX 924108TR
AssayComplete™ Thawing Reagent T8 is a reagent used to thaw cryopreserved DiscoverX cell lines. This product is provided as a bottle of 100mL.
AssayComplete™ Thawing Reagent T8 is an optimized thawing reagent for revival and recovery of DiscoverX cell lines. This reagent ensures optimal post-thaw viability, recovery and assay performance upon thawing. This product is provided as a bottle of 100mL. Please refer to cell line specific datasheet to determine the recommended thawing reagent.
Specifications
For Use With (Application) Cell Thawing Reagents
Quantity 100 mL