Learn More
Saint-Gobain AstiPure™ TP1 Series PFA Union Tee Fittings
Brand: Saint-Gobain TP10608
Additional Details : Weight : 0.03600kg
Specifications
Union Tee Fittings | |
PFA | |
For 6 x 8mm dia. tubing | |
45 mm |
5 mm | |
71 mm | |
1 Each |