Learn More
ASTM D2622/D7039, ECA Marine Fuel Curve, VHG™
Product Code. 17961196
Click to view available options
Quantity:
6 x 100 mL
Unit Size:
600mL
Product Code. 17961196
Brand: VHG Labs ALKKTS20MINW6MAR
VHG Labs offers ISO 17034 metallo-organic and aqueous inorganic certified reference materials for oil, fuel, and coolant analysis as well as a variety of certified standards for environmental, pharmaceutical, and industrial applications
ASTM D2622/D7039, ECA Marine Fuel Curve
TRUSTED_SUSTAINABILITY
Specifications
Shipping Condition Room Temperature
Certifications/Compliance ISO 17034
Solvent or Matrix 20 cSt Mineral Oil
Quantity 6 x 100 mL