missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ASTM Method D7796 Impurities in ETBE 0.1 - 93 Wt %, Dr. Ehrenstorfer
Specifications
Specifications
| Chemical Name or Material | 2-Ethoxybutane∣Diisobutylene∣Ethanol∣Ethyl tert-Butyl Ether (ETBE)∣Isobutene∣Methanol∣Methyl tert-butyl ether∣tert-Amyl Methyl Ether (TAME) |
| CAS | 2679-87-0, 25167-70-8, 64-17-5, 637-92-3, 115-11-7, 67-56-1 |
| Analyte or Component Names | 2-Ethoxybutane∣Diisobutylene∣Ethanol∣Ethyl tert-Butyl Ether (ETBE)∣Isobutene∣Methanol∣Methyl tert-butyl ether∣tert-Amyl Methyl Ether (TAME) |
| Shipping Condition | Room Temperature |
| Certifications/Compliance | ISO 17034 |
| Solvent or Matrix | ETBE |
| Industry Type | Food Testing |
| Quantity | 10 x 2 mL |
| Concentration or Composition (by Analyte or Components) | Methanol 0.09836 Wt %∣2-Ethoxybutane 0.492 %∣Diisobutylene 0.4924 Wt %∣Isobutene 0.4925 Wt %∣tert-Amyl Methyl Ether (TAME) 0.9862 Wt %∣Ethanol 0.9908 Wt %∣Methyl tert-butyl ether 2.959 Wt %∣Ethyl tert-Butyl Ether (ETBE) 92.89 Wt % |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?