Learn More
Astragaloside IV, Thermo Scientific™
Shop All Thermo Scientific Chemicals Products
Click to view available options
Quantity:
250 mg
1 g
Unit Size:
1g
250mg
Specifications
Specifications
| Quantity | 250 mg |
| Formula Weight | 784.97 |
| MDL Number | MFCD16036240,MFCD16036240 |
| Physical Form | Powder |
| Color | White to Almost-white |
| Purity | ≥97.5% |
| InChI Key | QMNWISYXSJWHRY-CSXKERSZSA-N |
| SMILES | CC(C)(O)[C@@H]1CC[C@@](C)(O1)[C@H]1[C@H](O)C[C@@]2(C)C3C[C@@H](O[C@@H]4O[C@H](CO)[C@@H](O)[C@H](O)[C@H]4O)C4[C@]5(CC35CC[C@]12C)CCC(O[C@@H]1OC[C@@H](O)[C@H](O)[C@H]1O)C4(C)C |
| IUPAC Name | (2R,3R,4S,5S,6R)-2-{[(3R,9R,12S,14R,15R,16R)-14-hydroxy-15-[(2R,5S)-5-(2-hydroxypropan-2-yl)-2-methyloxolan-2-yl]-7,7,12,16-tetramethyl-6-{[(2S,3R,4S,5R)-3,4,5-trihydroxyoxan-2-yl]oxy}pentacyclo[9.7.0.0¹,³.0³,⁸.0¹²,¹⁶]octadecan-9-yl]oxy}-6-(hydroxymethyl)oxane-3,4,5-triol |
| Molecular Weight (g/mol) | 784.98 |
| Show More |