Learn More
Invitrogen™ Astrovirus Capsid Monoclonal Antibody (4988)

Mouse Monoclonal Antibody
Brand: Invitrogen™ MA547548
Description
Specificity: Specific for the capsid protein. Pairs in ELISA to detect type 1. Other types not tested. Best ELISA pairs (capture/conjugate): AstrAstrovirus Capsid Monoclonal antibody (Product # MA5-47548)/Astrovirus Capsid Monoclonal antibody (Product # MA5-47546); Astrovirus Capsid Monoclonal antibody (Product # MA5-47548)/Astrovirus Capsid Monoclonal antibody (Product # MA5-47547). Purified preparations consist of >90% pure mouse monoclonal antibody which has been purified from ascites fluid by protein A chromatography. The final preparation is formulated to a protein concentration of 100 μg/mL in 0.01 M phosphate buffered saline, pH 7.2 and contains 0.1% sodium azide. Each vial contains 1.0 mL. This product contains no stabilizing proteins and should be stored at 4C until ready for use. Working dilution must be determined by the user. Suggested starting ranges are 1:10-1:50 for ICC/IF and 1:20-1:200. for ELISA.
The astrovirus capsid is the target of host antibodies including virus-neutralizing antibodies. The capsid protein also mediates the binding of host complement proteins and inhibits complement activation. [PMID: 28106836].
Specifications
| Astrovirus Capsid | |
| Monoclonal | |
| 100 μg/mL | |
| PBS with 0.1% sodium azide; pH 7.2 | |
| Proprietary sequence. | |
| 100 μg | |
| Primary | |
| 4°C | |
| Liquid |
| ELISA | |
| 4988 | |
| Unconjugated | |
| Mouse | |
| Protein A | |
| RUO | |
| Virus | |
| Antibody | |
| IgG2a |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.