Learn More
AT7867, MedChemExpress
Product Code. 16494958
Encompass_Preferred
Click to view available options
Quantity:
5 mg
10 mg
50 mg
100 mg
Unit Size:
100mg
10mg
50mg
5mg
Product Code. 16494958
Brand: Medchem Express HY12059
MedChemExpress AT7867 is a potent ATP-competitive inhibitor of Akt1/Akt2/Akt3 and p70S6K/PKA with IC50s of 32 nM/17 nM/47 nM and 85 nM/20 nM, respectively.
AT7867 is a potent ATP-competitive inhibitor of Akt1/Akt2/Akt3 and p70S6K/PKA with IC50s of 32 nM/17 nM/47 nM and 85 nM/20 nM, respectively.
Specifications
Chemical Name or Material AT7867
CAS 857531-00-1
Color White
Purity Grade Notes Research
Molecular Formula C20H20ClN3
Quantity 10 mg
Solubility Information DMSO : 10 mg/mL (29.60 mM; Need ultrasonic)
SMILES ClC1=CC=C(C=C1)C2(CCNCC2)C3=CC=C(C=C3)C4=CNN=C4
Molecular Weight (g/mol) 337.85
Formula Weight 337.85
Percent Purity 95.0%
Grade Research
Physical Form Solid
For Use With (Application) Cancer-Kinase/protease
Show More Show Less
Research purposes only