missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Ataxin 1 Antibody (S65-37), Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-42204
This item is not returnable.
View return policy
Description
Ataxin 1 Monoclonal antibody specifically detects Ataxin 1 in Human, Mouse, Rat samples. It is validated for Western Blot, Immunocytochemistry/ Immunofluorescence
Specifications
| Ataxin 1 | |
| Monoclonal | |
| 1 mg/mL | |
| Western Blot 1:1000, Immunocytochemistry/ Immunofluorescence | |
| P54254 | |
| Mouse | |
| Protein G purified | |
| RUO | |
| 6310 | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. | |
| IgG1 |
| Western Blot, Immunofluorescence | |
| S65-37 | |
| Unconjugated | |
| PBS (pH 7.4), 50% Glycerol | |
| ataxin 1, ataxin 1), ataxin-1, ATX1spinocerebellar ataxia 1 (olivopontocerebellar ataxia 1, autosomal dominant, SCA1D6S504E, Spinocerebellar ataxia type 1 protein | |
| Synthetic peptide amino acids 746-761 (RKRRWSAPETRKLEKS) of mouse ataxin-1. Rat: 93% identity (15/16 amino acids identical). Human: 87% identity (14/16 amino acids identical). | |
| 0.1 mg | |
| Primary | |
| Human, Mouse, Rat | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction