missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Novus Biologicals™ Ataxin-2 Antibody (05) [PE], Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals™ NBP3-26904PE
This item is not returnable.
View return policy
Description
Ataxin-2 Monoclonal antibody specifically detects Ataxin-2 in Human samples. It is validated for ELISA
Specifications
| Ataxin-2 | |
| Monoclonal | |
| PE | |
| ataxin 2, ataxin 2), ataxin-2, ATX2spinocerebellar ataxia 2 (olivopontocerebellar ataxia 2, autosomal dominant, SCA2FLJ46772, Spinocerebellar ataxia type 2 protein, TNRC13, trinucleotide repeat containing 13, Trinucleotide repeat-containing gene 13 protein | |
| This antibody was produced from a hybridoma resulting from the fusion of a mouse myeloma with B cells obtained from a mouse immunized with purified, recombinant Human Ataxin-2 (Uniprot#: Q99700; aa 733-942). | |
| 0.1 mL | |
| Cellular Markers, Hematopoietic Stem Cell Markers, Stem Cell Markers | |
| 6311 | |
| Store at 4°C in the dark. | |
| IgG |
| ELISA | |
| 05 | |
| PBS | |
| Mouse | |
| Protein A purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction