missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Ataxin-2 Antibody, DyLight 488, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NB100-58797G
This item is not returnable.
View return policy
Description
Ataxin-2 Polyclonal antibody specifically detects Ataxin-2 in Human, Mouse samples. It is validated for Western Blot
Specifications
| Ataxin-2 | |
| Polyclonal | |
| Western Blot | |
| ataxin 2, ataxin 2), ataxin-2, ATX2spinocerebellar ataxia 2 (olivopontocerebellar ataxia 2, autosomal dominant, SCA2FLJ46772, Spinocerebellar ataxia type 2 protein, TNRC13, trinucleotide repeat containing 13, Trinucleotide repeat-containing gene 13 protein | |
| The immunogen recognized by this antibody maps to a region between residue 1262 and 1312 of human ataxin 2 (spinocerebellar ataxia 2) using the numbering given in entry NP_002964.2 (GeneID 6311). | |
| 0.1 mL | |
| Cellular Markers, Hematopoietic Stem Cell Markers, Stem Cell Markers | |
| 6311 | |
| Store at 4C in the dark. | |
| IgG |
| Western Blot | |
| DyLight 488 | |
| 50mM Sodium Borate | |
| Rabbit | |
| Immunogen affinity purified | |
| RUO | |
| Primary | |
| Human, Mouse | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction