Learn More
Ataxin 2 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand:  Proteintech 21776-1-AP-20UL
 View more versions of this product
Product Code. 16851375
  • € 145.00 / 20µL
Description
The autosomal domit cerebellar ataxias (ADCA) are a heterogeneous group of neurodegenerative disorders characterized by progressive degeneration of the cerebellum, brain stem and spinal cord. Clinically, ADCA has been divided into three groups: ADCA types I-III. Defects in this gene are the cause of spinocerebellar ataxia type 2 (SCA2). SCA2 belongs to the autosomal domit cerebellar ataxias type I (ADCA I) which are characterized by cerebellar ataxia in combination with additional clinical features like optic atrophy, ophthalmoplegia, bulbar and extrapyramidal signs, peripheral neuropathy and dementia. SCA2 is caused by expansion of a CAG repeat in the coding region of this gene. Longer expansions result in earlier onset of the disease. Alternatively spliced transcript variants encoding different isoforms have been identified but their full length sequence has not been determined.
Specifications
Specifications
Ataxin 2
Polyclonal
Unconjugated
ATXN2
ataxin 2, ATX2, ATXN2, SCA2, TNRC13
Rabbit
Antigen Affinity Chromatography
RUO
20239, 288663, 6311
-20°C
Liquid
Immunofluorescence, Western Blot, Immunocytochemistry, Immunoprecipitation, Immunohistochemistry (Paraffin)
0.25 mg/mL
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
O70305, Q99700
Atxn2
Ataxin 2 Fusion Protein Ag16470
20 μL
Primary
Mouse, Human, Rat