missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Ataxin 2 Recombinant Rabbit Monoclonal Antibody (HL2885)

Rabbit Recombinant Monoclonal Antibody
Brand: Invitrogen™ MA556603
This item is not returnable.
View return policy
Description
Ataxin 2 Recombinant Monoclonal Antibody for ICC/IF, IHC (P), Western Blot
The autosomal dominant cerebellar ataxias (ADCA) are a heterogeneous group of neurodegenerative disorders characterized by progressive degeneration of the cerebellum, brain stem and spinal cord. Clinically, ADCA has been divided into three groups: ADCA types I-III. Defects in this gene are the cause of spinocerebellar ataxia type 2 (SCA2). SCA2 belongs to the autosomal dominant cerebellar ataxias type I (ADCA I) which are characterized by cerebellar ataxia in combination with additional clinical features like optic atrophy, ophthalmoplegia, bulbar and extrapyramidal signs, peripheral neuropathy and dementia. SCA2 is caused by expansion of a CAG repeat in the coding region of this gene. Longer expansions result in earlier onset of the disease. Alternatively spliced transcript variants encoding different isoforms have been identified but their full length sequence has not been determined.
Specifications
| Ataxin 2 | |
| Recombinant Monoclonal | |
| 1 mg/mL | |
| PBS with no preservative | |
| Q99700 | |
| Atxn2 | |
| Recombinant fragment of human ATXN2. | |
| 100 μL | |
| Primary | |
| Human, Rat | |
| Antibody | |
| IgG |
| Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry | |
| HL2885 | |
| Unconjugated | |
| Atxn2 | |
| 9630045M23Rik; ASL13; ataxin 2; ataxin-2; ATX2; Atxn2; AW544490; FLJ46772; SCA2; spinocerebellar ataxia 2 (olivopontocerebellar ataxia 2, autosomal dominant, ataxin 2); spinocerebellar ataxia 2 homolog; spinocerebellar ataxia type 2 protein; Spinocerebellar ataxia type 2 protein homolog; TNRC13; Trinucleotide repeat-containing gene 13 protein | |
| Rabbit | |
| Protein A | |
| RUO | |
| 288663, 6311 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction