Learn More
Ataxin-3 Antibody (S04-1I8), Novus Biologicals™
Rabbit Monoclonal Antibody
Brand: Novus Biologicals NBP3-19834-100ul
This item is not returnable.
View return policy
Description
Ataxin-3 Monoclonal antibody specifically detects Ataxin-3 in Human, Mouse, Rat samples. It is validated for Western Blot
Specifications
| Ataxin-3 | |
| Monoclonal | |
| 0.3 mg/mL | |
| 50mM Tris-Glycine(pH 7.4), 0.15M NaCl, 40% Glycerol, 0.05% BSA | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| Primary | |
| Human, Mouse, Rat | |
| Purified |
| Western Blot | |
| S04-1I8 | |
| Unconjugated | |
| ataxin 3, ATX3AT3, EC 3.4.19.12, EC 3.4.22, Machado-Joseph disease protein 1, MJD1, MJDolivopontocerebellar ataxia3, autosomal dominant, ataxin 3), olivopontocerebellar ataxia 3, Spinocerebellar ataxia type 3 protein | |
| A synthetic peptide of human Ataxin-3 (Uniprot # P54252) | |
| 100 μg | |
| Cell Biology, Neurodegeneration, Neuroscience | |
| 4287 | |
| Store at 4°C short term. Store at -20°C long term. Avoid freeze-thaw cycles. | |
| IgG |