Learn More
Ataxin-3 Antibody (S04-1I8), Novus Biologicals™
Rabbit Monoclonal Antibody
Brand:  Novus Biologicals NBP3-19834-100ul
 View more versions of this product
Product Code. 18697663
  • € 645.00 / 100µL
Description
Ataxin-3 Monoclonal antibody specifically detects Ataxin-3 in Human, Mouse, Rat samples. It is validated for Western Blot
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Ataxin-3
Monoclonal
0.3 mg/mL
50mM Tris-Glycine(pH 7.4), 0.15M NaCl, 40% Glycerol, 0.05% BSA
Rabbit
Affinity purified
RUO
Primary
Human, Mouse, Rat
Purified
Western Blot
S04-1I8
Unconjugated
ataxin 3, ATX3AT3, EC 3.4.19.12, EC 3.4.22, Machado-Joseph disease protein 1, MJD1, MJDolivopontocerebellar ataxia3, autosomal dominant, ataxin 3), olivopontocerebellar ataxia 3, Spinocerebellar ataxia type 3 protein
A synthetic peptide of human Ataxin-3 (Uniprot # P54252)
100 μg
Cell Biology, Neurodegeneration, Neuroscience
4287
Store at 4°C short term. Store at -20°C long term. Avoid freeze-thaw cycles.